Learn More
Bullard™ Dual-Cool™ Climate Control Assemblies, Quick-Disconnect Nipples
Shop All Bullard Company Products
Specifications
Specifications
| Certifications/Compliance | CE, NIOSH |
| For Use With (Equipment) | DC Vests of GenVX, 88VX, CC20 and GR50 Series Airline Respirator |
| Material | Snap-tite Brass |
| Includes | CH60 Connector Hose and Nylon Belt |
| Model | DC5043 |
| Size | 0.25 in. |
| Product Type | Climate Control Device |