Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ HEPA Filter and Combination Cartridges for EVA™ Powered Air Purifying Respirator
SDP

Catalog No. 19808700 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
HEPA Filter Cartridge
HEPA/Chemical Cartridge
HEPA/OV/Chemical Cartridge
Color:
Magenta
Magenta/Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
19-808-700 HEPA/Chemical Cartridge -
19-824-560 HEPA/OV/Chemical Cartridge Magenta/Yellow
17-986-282 HEPA Filter Cartridge Magenta
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-808-700 Supplier Bullard™ Supplier No. PAPRFC5
Add to Cart
Add to Cart

Filters and combination cartridges are designed for use with EVA series PAPR systems and provide protection against particulates.

  • Protection: Particulates, Ammonia, Formaldehyde, Methylamine, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride
  • Particulate Efficiency: High Efficiency Particulate Absolute (HEPA); 99.99% vs. a mass mean 0.3 micron aerosol at 85 LPM; Media secured with urethane glue around perimeter
  • Filter Housing Material: ABS Plastic
  • Sorbent Bed: Activated Carbon (Chromium Free)
  • Regulatory Approvals: USA; NIOSH 42 CFR Part 84; TC-23C-2886 (EVA PAPR with Loose Fitting Headtops) - per test procedures: TEB-APR-STP-0001, RCT-APR-STP-0035, RCT-APR-STP-0036, RCT-APR-STP-0040, RCT-APR-STP-0042, RCT-APR-STP-0046, RCT-APR-STP-0048
  • Packaging: 6 per box; Each filter individually sealed in plastic
  • Lot Code Identification: Yes
  • Expiration/Shelf Life: 5 years (unopened with proper storage conditions)

Compliance

Tested in accordance to NIOSH 42 CFR Part 84; EN 12941; CE 1094 (EVA PAPR with Loose Fitting Headtops only) requirements.

Specifications

Filter Type Combination Mechanical and Chemical
Product Type HEPA/Chemical Cartridge
For Use With (Equipment) Eva™ powered air purifying respirator

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.