Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ EVA™ Powered-Air Purifying Respirator Shower Caps
SDP

Catalog No. 19830081 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
EVA™ FC1 and FC2 PAPR System
EVA™ FC3 PAPR System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-830-081 EVA™ FC1 and FC2 PAPR System
17-986-283 EVA™ FC3 PAPR System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-830-081 Supplier Bullard™ Supplier No. PAPRSC1
This item is not returnable. View return policy

Shower cap accessories and replacement parts for EVA™ PAPR systems.

  • Parts for EVA™ PAPR unit
  • Shower cap accessories

Specifications

Color White
Disposable Reusable
Product Type Shower Cap
For Use With (Application) For Decon Shower
For Use With (Equipment) EVA™ FC1 and FC2 PAPR System
Model PAPRSC1

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.