Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ EVA™ PAPR Breathing Tube Covers
SDP

Catalog No. 19065764 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Polypropylene
Tychem SL
Color:
Clear
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color
19-065-764 Tychem SL White
19-824-424 Polypropylene Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-065-764 Supplier Bullard™ Supplier No. 20SLBTC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Disposable breathing tube covers keep the EVA™ PAPR clean from debris.

Specifications

Color White
Material Tychem SL
Product Type Breathing Tube Cover
For Use With (Equipment) EVA™ PAPR Systems
Model 20SLBTC
Length (English) 24.5 in.
Length (Metric) 62.2 cm

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.