Learn More
Bullard™ EVAHL PAPR System For Use with Hazardous Locations
SDP
Narrow profile PAPR offers workers protection in hazardous locations. System includes loose-fitting facepiece, blower, belt, battery, breathing tube, charger, air flow indicator, and head top with ratchet suspension.
Supplier:  Bullard™ EVAHL20LFMDB
 View more versions of this product
Catalog No. 17-989-506
Description
Description
  • HazLoc rated†
  • For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
  • Class II, Division 2, Groups F, G (Dusts)
  • Class III (Fibers or Flyings)
  • Simple, one button operation
  • Choice of two air speeds
  • Single filter design
  • Active Flow Technology responds to air flow conditions
  • 25 Assigned Protection Factor (APF) for Loose Fitting Headtops
  • Independent, 3rd Party Documentation for Assigned Protection Factor of 1000 for certain style Hoods (RT, CC20) and Spectrum Full face mask
  • Dual alarms for low flow and low battery
  • Visual fuel gauge
  • 10X the life of most brush motors
  • Genius Charger compatible
†Not for use in Oxygen deficient atmospheres
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.
Specifications
Specifications
ANSI/ISA 2.12.01-2007
Lithium-ion Polymer
NIOSH
PAPR System with Facepiece
8.5/7 CFM
Decon Belt
7 to 8 hours
Vinyl
CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
Medium
Painting, chemical handling, healthcare, pharmaceutical
EVAHL20LFMDB
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.