Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ EVAHL PAPR System For Use with Hazardous Locations
SDP

Catalog No. 17989508 Shop All Bullard Company Products
Change view
Click to view available options
Size:
Large (Narrow)
Medium
Medium (Narrow)
Small (Narrow)
Belt Material:
Foam
Vinyl
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Belt Material
17-989-508 Medium (Narrow) Vinyl
17-989-500 - Foam
17-989-503 Medium (Narrow) Foam
17-989-502 Large (Narrow) Foam
17-989-506 Medium Vinyl
17-989-505 - Vinyl
17-989-504 Small (Narrow) Foam
17-989-509 Small (Narrow) Vinyl
17-989-507 Large (Narrow) Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-989-508 Supplier Bullard™ Supplier No. EVAHL20LF2MDB

Narrow profile PAPR offers workers protection in hazardous locations. System includes loose-fitting facepiece, blower, belt, battery, breathing tube, charger, air flow indicator, and head top with ratchet suspension.

  • HazLoc rated†
  • For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
  • Class II, Division 2, Groups F, G (Dusts)
  • Class III (Fibers or Flyings)
  • Simple, one button operation
  • Choice of two air speeds
  • Single filter design
  • Active Flow Technology responds to air flow conditions
  • 25 Assigned Protection Factor (APF) for Loose Fitting Headtops
  • Independent, 3rd Party Documentation for Assigned Protection Factor of 1000 for certain style Hoods (RT, CC20) and Spectrum Full face mask
  • Dual alarms for low flow and low battery
  • Visual fuel gauge
  • 10X the life of most brush motors
  • Genius Charger compatible
†Not for use in Oxygen deficient atmospheres

Compliance

Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.

†CSA International Certificate 2510928 per ANSI/ISA 12.12.01-2007

Specifications

ANSI Standard ANSI/ISA 2.12.01-2007
Battery Life 7 to 8 hours
Battery Type Lithium-ion Polymer
Belt Material Vinyl
Certifications/Compliance NIOSH
Certifications (CSA) CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
Product Type PAPR System with Facepiece
Size Medium (Narrow)
Flow Rate 8.5/7 CFM
For Use With (Application) Painting, chemical handling, healthcare, pharmaceutical
Includes Decon Belt
Model EVAHL20LF2MDB
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.