Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, Double Bib Hood
Shop All Bullard Company Products
Description
- Ideal for painting, chemical handling, healthcare, pharmaceutical applications
- Choice of hood configurations
- Airflow rate: 8.5/7 cfm
- 77dBA low voltage and 77dBA low flow alarm
- Lithium ion polymer battery (1-year life span)
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Specifications
| Battery Type | Lithium-ion Polymer |
| Certifications/Compliance | ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III |
| Product Type | Double Bib Suspension-free Hood |
| For Use With (Application) | Ideal for painting, chemical handling, healthcare, pharmaceutical |
| Includes | Blower, belt, battery, breathing tube, headtop, charger, inflatable cuff |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.