Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, Double Bib Hood
SDP
Catalog No. 17989533 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Double Bib Hood
Double Bib Hood with Vinyl Decon Belt
Double Bib Saranex™ Hood
Double Bib Saranex™ Hood with Vinyl Decon Belt
Double Bib Suspension-free Hood
Double Bib Suspension-free Hood with Vinyl Decon Belt
For Use With (Application):
Ideal for painting, chemical handling, healthcare, pharmaceutical
Painting, chemical handling
Includes:
Blower, belt, battery, breathing tube, headtop, charger, inflatable cuff
Blower, belt, battery, breathing tube, headtop, charger, vinyl decon belt
Blower, vinyl decon belt, battery, breathing tube, headtop, charger, inflatable cuff
Ratchet suspension
Ratchet suspension, Vinyl decon belt
S51 hard hat, ratchet suspension, blower, belt, battery, breathing tube, headtop & charger
Saranex™ Hood, Ratchet suspension
Saranex™ Hood, ratchet suspension and vinyl decon belt
Sport cuff, blower, belt, battery, breathing tube, headtop & charger
Sport cuff, blower, belt, battery, breathing tube, headtop, charger, vinyl decon belt
Suspension Type:
Ratchet suspension
Suspension-free
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application) Includes Suspension Type
17-989-533 Double Bib Suspension-free Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical Sport cuff, blower, belt, battery, breathing tube, headtop, charger, vinyl decon belt Suspension-free
17-989-510 Double Bib Hood Painting, chemical handling Ratchet suspension Ratchet suspension
17-989-512 Double Bib Saranex™ Hood - Saranex™ Hood, Ratchet suspension Ratchet suspension
17-989-514 Double Bib Hood - S51 hard hat, ratchet suspension, blower, belt, battery, breathing tube, headtop & charger Ratchet suspension
17-989-531 Double Bib Suspension-free Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical - -
17-989-530 Double Bib Suspension-free Hood - - Suspension-free
17-989-511 Double Bib Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical Ratchet suspension, Vinyl decon belt Ratchet suspension
17-989-513 Double Bib Saranex™ Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical Saranex™ Hood, ratchet suspension and vinyl decon belt Ratchet suspension
17-989-515 Double Bib Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical Blower, belt, battery, breathing tube, headtop, charger, vinyl decon belt -
17-989-528 Double Bib Suspension-free Hood Ideal for painting, chemical handling, healthcare, pharmaceutical Blower, belt, battery, breathing tube, headtop, charger, inflatable cuff -
17-989-529 Double Bib Suspension-free Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical Blower, vinyl decon belt, battery, breathing tube, headtop, charger, inflatable cuff -
17-989-532 Double Bib Suspension-free Hood - Sport cuff, blower, belt, battery, breathing tube, headtop & charger Suspension-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 17-989-533 Supplier Bullard™ Supplier No. EVAHLRT4TDB
Designed to be used with the EVAHL Powered Air Purifying Respirator
  • Ideal for painting, chemical handling, healthcare, pharmaceutical applications
  • Choice of hood configurations
  • Airflow rate: 8.5/7 cfm
  • 77dBA low voltage and 77dBA low flow alarm
  • Lithium ion polymer battery (1-year life span)
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Battery Type Lithium-ion Polymer
Certifications/Compliance ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Product Type Double Bib Suspension-free Hood with Vinyl Decon Belt
For Use With (Application) Ideal for painting, chemical handling, healthcare, pharmaceutical
Includes Sport cuff, blower, belt, battery, breathing tube, headtop, charger, vinyl decon belt
Suspension Type Suspension-free
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.