Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, QC Hood
Shop All Bullard Company Products
Description
- Dupont™ Tychem QC hood
- Double bib with ratchet suspension
- Sealed seams (Cat. No. 17989516 only)
- Threaded connector style
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Specifications
| Battery Type | Lithium-ion Polymer |
| Certifications/Compliance | ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III |
| Connection Type | Clamp Connector Style/Threaded Connector Style |
| Product Type | Double Bib Hood |
| For Use With (Application) | Ideal for painting, chemical handling, healthcare, pharmaceutical |
| Includes | Solvent Resistant Lens |
| Suspension Type | Ratchet suspension |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.