Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, QC Hood
SDP

Catalog No. 17989525 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Double Bib Hood
Single Bib Hood
Includes:
Oversized Lens, Inflatable Neck Cuff
Ratchet Suspension
Solvent Resistant Lens
Vinyl Decon Belt
Vinyl Decon Belt, Ratchet Suspension
Vinyl Decon Belt, Sealed Seams
Vinyl Decon Belt, Solvent Resistant Lens
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
17-989-525 Single Bib Hood Vinyl Decon Belt, Solvent Resistant Lens
17-989-519 Double Bib Hood Vinyl Decon Belt, Ratchet Suspension
17-989-518 Double Bib Hood Ratchet Suspension
17-989-520 Single Bib Hood Ratchet Suspension
17-989-521 Single Bib Hood Vinyl Decon Belt
17-989-517 Double Bib Hood Vinyl Decon Belt, Sealed Seams
17-989-522 Double Bib Hood Solvent Resistant Lens
17-989-523 Double Bib Hood Vinyl Decon Belt
17-989-526 Double Bib Hood Oversized Lens, Inflatable Neck Cuff
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-989-525 Supplier Bullard™ Supplier No. EVAHL20TPTDB

Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator

  • Dupont™ Tychem QC hood
  • Double bib with ratchet suspension
  • Sealed seams (Cat. No. 17989516 only)
  • Threaded connector style

Certifications

ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III

Specifications

Battery Type Lithium-ion Polymer
Certifications/Compliance ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Product Type Single Bib Hood
For Use With (Application) Ideal for painting, chemical handling, healthcare, pharmaceutical
Includes Vinyl Decon Belt, Solvent Resistant Lens
Suspension Type Ratchet suspension

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.