Learn More
Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Shop All Bullard Company Products
Description
- Loose fitting facepiece
- Head covers made of Tychem
- Sewn-in headband suspension
- Twist-lock breathing tube connection
- For use with PAPR and SAR systems
- Choose from a variety of sizes and color connections
- Acetate lens material
- NIOSH approved
Specifications
Specifications
| Assigned Protection Factor (APF) | 25 |
| Product Type | Loose Fitting Facepiece |
| Size | Medium |
| For Use With (Application) | Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare |
| For Use With (Equipment) | Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator |
| Includes | Contoured-fit, blue twist lock connection |
| Latex Free | Yes |
| Model | 20LF2M50 |
| Packaging Type | Bulk Pack |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.