Learn More
Description
- Clear visors accommodate safety spectacles, goggles, and respirators to protect face from numerous industrial hazards
- Flat acetate visors are for chemical applications and light impact protection
- Heat-formed polycarbonate visors are for high-heat operations with little or no chemical exposure, and are curved for easy attachment and reduced stress cracking
- Visors are 15 in. wide (20.3cm) and available in 6, 8, and 10 in. lengths (15.2, 20.3, 25.4cm)
Order Info
Do not use any faceshield unless suitable primary eye protection is used.
Compliance
Meets ANSI/ISEA Z87.1 and ANSI Z87 standards.
Specifications
Specifications
| Dimensions (L x W) | 8 x 15 in. |
| Width (Metric) | 38.1 cm |
| Shape | Flat |
| Thickness (English) | 0.04 in. |
| Thickness (Metric) | 1.0 cm |
| Width (English) | 15 in. |
| Certifications/Compliance | ANSI Z87.1 |
| Lens Tint | Clear |
| Product Type | Clear flat acetate visor |
| Length (English) | 8 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
