Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Faceshield Visors
SDP

Catalog No. 19021443 Shop All Bullard Company Products
Change view
Click to view available options
Lens Material:
Acetate
Polycarbonate
Dimensions (L x W):
6 x 15 in.
7 x 15 in.
8 x 15 in.
10 x 15 in.
Thickness (English):
0.04 in.
0.040 in.
0.060 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Material Dimensions (L x W) Thickness (English)
19-021443 Acetate 6 x 15 in. 0.040 in.
19-021321 Acetate 10 x 15 in. 0.040 in.
19-021464 Polycarbonate 8 x 15 in. 0.040 in.
19-021466 Polycarbonate 8 x 15 in. 0.060 in.
17-361 Acetate 8 x 15 in. 0.04 in.
19-088-776B Polycarbonate 7 x 15 in. 0.040 in.
19-088-776A Acetate 7 x 15 in. 0.040 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-021443 Supplier Bullard™ Supplier No. 640
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with Bullard headgear

  • Clear visors accommodate safety spectacles, goggles, and respirators to protect face from numerous industrial hazards
  • Flat acetate visors are for chemical applications and light impact protection
  • Heat-formed polycarbonate visors are for high-heat operations with little or no chemical exposure, and are curved for easy attachment and reduced stress cracking
  • Visors are 15 in. wide (20.3cm) and available in 6, 8, and 10 in. lengths (15.2, 20.3, 25.4cm)

Order Info

Do not use any faceshield unless suitable primary eye protection is used.

Compliance

Meets ANSI/ISEA Z87.1 and ANSI Z87 standards.

Specifications

Dimensions (L x W) 6 x 15 in.
Thickness (English) 0.040 in.
Shape Flat
Thickness (Metric) 1.0 cm
Width (English) 15 in.
Width (Metric) 38.1 cm
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Clear flat acetate visor
Length (English) 6 in.
Length (Metric) 15.2 cm
Lens Material Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.