Learn More
Bullard™ Constant Flow Assemblies for RT Series Airline Respirators
Shop All Bullard Company Products
Description
- Constant flow accessory
- For use with RT series airline respirator
- Flow control assemblies
- Nipple types include bayonet, CEJN, industrial interchange, schrader, or snap-tite
- Does not include breathing tube
Specifications
Specifications
| Connection Type | 0.25 in. Snap-Tite Stainless Steel Nipple |
| Controller Type | Constant Flow |
| Material | Stainless Steel |
| Product Type | Flow Control Assembly |
| For Use With (Equipment) | RT Series Airline Respirator |
| Model | F34 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.