Learn More
Bullard™ Free-Air™ ADP20 Air Driven Pump Accessory, Service Kit
SDP
Catalog No. 17604219 Shop All Bullard Company Products
Change view
Click to view available options
Service Kit
Catalog No.
17-604-219 Service Kit
1 options
Catalog No. 17-604-219 Supplier Bullard™ Supplier No. 17761
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Service kit accessory for use with ADP20 air driven pump.
Kit Components
  • 4 Vanes
  • 2 Motor bearings
  • 4 Vane springs
  • Seal
  • 2 Body gaskets
  • 2 Push pins
  • End cap gasket and o-ring
  • Instructions
Specifications
Model 17761
Product Type Service Kit