Learn More
Bullard™ EDP16TE Free-Air™ Pump
SDP
Catalog No. 17989629 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Breathing Air Pump
Catalog No. Product Type
17-989-629 Breathing Air Pump
1 options
Catalog No. 17-989-629 Supplier Bullard™ Supplier No. EDP16TE
Portable, electric driven pumps with capacity to supply air to two hood-style or three half or full-face mask airline respirators.
  • Two to three person air pump
  • Oil-less eliminates OSHA required CO monitors, temperature alarms, airline filters
  • 1 1/2 horse power electric driven pump
  • Portable (includes handles)
  • Pressure gauge (0-30 psig)
  • Two 1/2 in. Industrial Interchange (Hansen compatible) quick-disconnect coupler
Warranty and Services
One year
Specifications
Model EDP16TE
Pressure 0 to 30 psi
Overall Length (English) 28 in.
Weight (Metric) 44.5 kg
Product Type Breathing Air Pump