Learn More
Bullard™ Free-Air EDP30 Electric Driven Pumps
SDP
Catalog No. 17604209 Shop All Bullard Company Products
Catalog No. 17-604-209 Supplier Bullard™ Supplier No. EDP30C460
Supports climate control devices
Shipped complete with inlet and outlet filter cartridges, pressure gauges, pressure relief valves, and one air supply hose quick-disconnect pod with six 0.5 in. Industrial Interchange (Hansen compatible) quick-disconnect couplers.
  • Supplies air for four hood-style, six half or full face mask airline respirators, or two hood-style respirators equipped with Frigitron climate control devices
  • Respirators must be approved to operate on 30 psig or less
  • Easy operation and low maintenance
  • Completely mobile and portable
  • No expensive installation costs
Specifications
Applications Support Climate Control Devices
Includes Inlet and outlet filter cartridge, pressure gauges, pressure relief valves and one air supply hose quick-disconnect pod with six 0.5 in. industrial interchange quick-disconnect couplers, reversing starter
Product Type Electric Driven Air Pump