Learn More
Bullard™ Free-Air EDP50 Electric Driven Pumps
SDP
Catalog No. 17604216 Shop All Bullard Company Products
Catalog No. 17-604-216 Supplier Bullard™ Supplier No. EDP50C460
Supports up to ten respirators
Operates without temperature alarms, CO monitors or airline filters.
  • Supplies air for seven hood-style or ten half or full face mask airline respirators
  • Features point of attachment, quick disconnect pods that can be extended up to 300 ft. from pump
  • Air flow of 50 cfm at 5 psig
  • Easy operation and low maintenance
  • Completely mobile and portable
  • No expensive installation costs
Specifications
Includes Inlet and outlet filter cartridge, pressure gauges, pressure relief valves and one air supply hose quick-disconnect pod with six 0.5 in. industrial interchange quick-disconnect couplers
Product Type Electric Driven Air Pump