Learn More
Bullard™ Free-Air™ EDP30 Electric Driven Pump Accessory, Service Kit
SDP
Catalog No. 17604218 Shop All Bullard Company Products
Change view
Click to view available options
:
Service kit for Electric Driven Pump 30
Catalog No.
17-604-218 Service kit for Electric Driven Pump 30
1 options
Catalog No. 17-604-218 Supplier Bullard™ Supplier No. 15924
Service kit accessory for use with EDP30 electric driven pump.
  • For Bullard Free-Air EDP30 Electric Driven Pump
  • Service Kit includes 4 carbon vanes, 2 bearings, and 2 cover gaskets
Specifications
For Use With (Equipment) Free-Air Pump
Includes 4 carbon vanes, 2 bearings and 2 cover gaskets
Model 15924
Product Type Service Kit