Learn More
Bullard™ CAB Series Remote Alarm for CO Monitor
SDP
Catalog No. 19100674 Shop All Bullard Company Products
Change view
Click to view available options
Model:
RA
Catalog No. Model
19-100-674 RA
1 options
This item is not returnable. View return policy
Catalog No. 19-100-674 Supplier Bullard™ Supplier No. RA
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remote alarm accessory for use with CAB series.
  • Accessory for Clean Air Boxes (CAB)
  • Safety mechanism for CO monitoring
  • Strobe light with siren
  • 50 ft. cord
Specifications
Model RA