Learn More
Bullard™ Six-Point Suspension Ratchet Caps
SDP
Catalog No. 19807753 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Hi-vis Orange
Navy Blue with AKZO Nobel Logo
Soft Green with AKZO Nobel Logo
White with AKZO Nobel Logo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-753 Navy Blue with AKZO Nobel Logo
19-807-755 Hi-vis Orange
19-807-751 White with AKZO Nobel Logo
19-807-752 Soft Green with AKZO Nobel Logo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-807-753 Supplier Bullard™ Supplier No. 30NBR1000W/BYB29822
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Classic cap style with comfortable six-point suspension.
  • Classic Series exceeds ANSI/ISEA Z89.1-2014, Type 1, Class E and G requirements, and meets CSA Z94.1-2005, Type I, Class E and G requirement
  • With AKZO Nobel logo
  • Available in different colors
Specifications
Certifications/Compliance Exceeds ANSI/ISEA Z89.1-2014, Type 1, Class E and G requirements, and meets CSA Z94.1-2005, Type I, Class E and G requirements
Color Navy Blue with AKZO Nobel Logo
Style Classic
Suspension Type Six-point
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.