Learn More
Bullard™ Spectrum™ Series Full-Facepiece Airline Respirators
SDP
Catalog No. 19034627 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
High Pressure Respirator Assembly
Low Pressure Respirator Assembly
For Use With (Equipment):
Breathing air compressor
Breathing air cylinder
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment) Size
19-034-627 High Pressure Respirator Assembly Breathing air compressor Medium/Large
19-034-628 Low Pressure Respirator Assembly Breathing air cylinder Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-627 Supplier Bullard™ Supplier No. SPEC40L
Continuous-flow respirators feature lightweight facepieces with a wide-angle lens for a clear field of view.
  • Continuous-flow, high-pressure model includes breathing tube and flow valve for compressor use
  • Continuous-flow, low-pressure model includes breathing tube and constant flow valve for air pump use
  • Silicone flange seals for comfortable fit
  • SnapFit™ design for easy donning
  • Clear silicone nosecup and low-profile regulator minimize interference with vision and other PPE
  • Regulator allows donning without free flow of air, conserving air supply
  • Swivel connector provides free movement of regulator hose for comfort and maneuverability
  • All models equipped with medium/large facepiece
  • Can be used with V10 and V20 Series breathing air hoses (order separately)
Compliance
NIOSH approved.
Specifications
Certifications/Compliance NIOSH
Product Type High Pressure Respirator Assembly
Size Medium/Large
For Use With (Equipment) Breathing air compressor
Includes Headstrap, breathing tube assembly with flow control device and nylon belt
Model SPEC40L
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.