Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Supplied-Air Respirator Accessories, Belts
SDP

Replacement belt features durable nylon or vinyl construction for use with belt-mounted SAR climate control devices.

$40.70 - $45.10

Specifications

For Use With (Equipment) 20LF Series Airline Respirators
Overall Length (English) 54 in.
Overall Length (Metric) 137 cm
Product Type Belt
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-059-266
SDP
Required Accessories View Documents
Bullard™
4612
Nylon
Each for $45.10
Only null left
Add to Cart
 
19-824-382
SDP
Required Accessories View Documents
Bullard™
36501
Vinyl Decon
Each for $40.70
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

20LF Series Airline Respirators
54 in.
137 cm
Belt
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.