Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ RT Series Supplied-Air Respirator Breathing Tubes
SDP

Catalog No. 19824425 Shop All Bullard Company Products
Change view
Click to view available options
Description:
Lightweight, disposable breathing tube
Wire-reinforced, heavy-duty breathing tube
Color:
Gray
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Color
19-824-425 Lightweight, disposable breathing tube Gray
EB-20BT Wire-reinforced, heavy-duty breathing tube White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-425 Supplier Bullard™ Supplier No. RTBT
Add to Cart
Add to Cart

Breathing tube component for use with RT Series respirators.

  • Component for use with RT series respirators
  • Choose between heavy-duty or lightweight options
  • Breathing tubes must be used in conjunction with hood, flow control device, and air hose

Specifications

Description Lightweight, disposable breathing tube
Color Gray
Product Type Breathing Tube
For Use With (Equipment) RT Series supplied-air respirators
Includes S18051 hose clamp
Model RTBT

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.