Description
- Loose fitting respirator double bib hoods
- Choose between ratchet or no suspension models
- Threaded connection for PAPR
- DuPont Tychem 2000 material construction
- 1,000 Assigned Protection Factor (APF)
Specifications
Specifications
| Assigned Protection Factor (APF) | 1000 |
| Connection Type | Clamp |
| Material | Tychem QC |
| Product Type | Double Bib Hood |
| For Use With (Application) | Pharmaceutical, Chemical, Pesticides, Spray Painting and Coating, Nuclear |
| For Use With (Equipment) | Supplied Air Respirator (CC20 Series) or Powered Air Respirator (EVA, EVAHL, PA20, PA30 or PA30IS Series) |
| Lens Type | Acetate |
| Model | 20TPCN |
| Suspension Type | No Suspension |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.