Description
- Connect the respirator’s breathing tube fitting to the female quick disconnect coupler
- choose from differet hose lengths and quick disconnect coupler options
- Extension hose kit includes:
- One V10 extension hose with female fittings on both ends
- One V11 3/8” hose-to-3/8” hose adaptor
- One V13 3/8” hose-to-3/8” pipe adaptor
Specifications
Specifications
| Connection Type | Schrader, Steel quick-disconnect Coupler |
| Color | Black |
| Product Type | Hose Kit |
| Includes | V10 starter hose, female quick-disconnect coupler, standard female hose thread, V13 hose-to-pipe adaptor |
| Model | 46913 |
| Length (English) | 25 ft. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.