Description
Provides impact protection from all directions: top, front, sides and rear
- 4-point suspension
- Replaceable, moisture wicking Sportek™ brow pad
- Built-in goggle strap retaining slots
- Polyethylene outer shell
- EPS foam impact liner
Order Info
Available in either 4-point Flex-Gear™ or self-sizing suspension
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z891-1997; Type 2; Class E G and C; CSA Z94.1-199 |
| Color | Hi-vis Orange |
| Suspension Type | Pin-Lock |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.