Learn More
Bullard™ Vector Cap-Style Helmet
SDP
Catalog No. 19098035 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Hi-vis Green
Hi-vis Orange
Hi-vis Yellow
Suspension Type:
Pin-Lock
Ratchet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-098-035 Hi-vis Yellow Pin-Lock
19-098-036 Hi-vis Green Ratchet
19-098-038 Hi-vis Yellow Ratchet
19-098-033 Hi-vis Green Pin-Lock
19-098-034 Hi-vis Orange Pin-Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-098-035 Supplier Bullard™ Supplier No. VTHYP
Complies with ANSI/ISEA Z89.1-2009 Type II, as well as CSA Z94.1-2005, Type II head protection standards
Provides impact protection from all directions: top, front, sides and rear
  • 4-point suspension
  • Replaceable, moisture wicking Sportek™ brow pad
  • Built-in goggle strap retaining slots
  • Polyethylene outer shell
  • EPS foam impact liner
Order Info
Available in either 4-point Flex-Gear™ or self-sizing suspension
Specifications
Certifications/Compliance Meets ANSI Z891-1997; Type 2; Class E G and C; CSA Z94.1-199
Color Hi-vis Yellow
Suspension Type Pin-Lock
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.