Learn More
Stoelting™ Bulldog Clamp
SDP
Stoelting™ Bulldog Clamp is useful in surgeries that require precise blood stoppage.
$176.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 3
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
10-001-372
SDP
View Documents
Stoelting™
5212037
1.5 in. Straight
Each for $176.00
Only null left
Add to Cart
10-001-371
SDP
View Documents
Stoelting™
5212036
2 in. Straight
Each for $176.00
Only null left
Add to Cart
10-001-370
SDP
View Documents
Stoelting™
5212035
2 in. Curved
Each for $176.00
Only null left
Add to Cart
Description
Description
  • One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped.
  • In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.