Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Work Shirt, Navy

Catalog No. 18120300 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
Large
Large (Long)
Medium
Medium (Long)
Small
X-Large
X-Large (Long)
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-120-300 X-Large
18-120-297 Medium (Long)
18-120-298 Large
18-120-299 Large (Long)
18-120-295 Small
18-120-296 Medium
18-120-301 X-Large (Long)
18-120-302 2X-Large
18-120-303 2X-Large (Long)
18-120-305 3X-Large (Long)
18-120-304 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-120-300 Supplier Bulwark™ FR Supplier No. SLW2NV RG XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Men's Midweight Excel FR™ Comfortouch™ Work Shirt is your go-to FR work shirt with CAT 2 AR/FR protection to keep you compliant.

  • Certifications Level 1: ASTM F1506, ANSI 107, NFPA 70E
  • Certifications Level 2: Category 2 Protection
  • Hazard: 2
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 88% Cotton/12% Nylon
  • Closure: Button-down front with one-piece lined cuffs with button closures
  • Collar: Lay-flat collar
  • Pocket: Two chest pockets with button flap closures and sewn-in pencil stall
  • Features: Hemmed front with button closures
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Size X-Large
Gender Men's
Material Cotton, Nylon
Color Navy
Closure Type Button
Hazard Risk Category 2
Arc Flash Rating ATPV 8.6 Calories/cm2
Certifications/Compliance ASTM F1506, ANSI 107, NFPA 70E, Category 2 Protection
Weight (English) 7 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.