Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Flame Resistant Dress Uniform Shirt

Catalog No. 19295123 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
X-Large
2X-Large
Size (Metric) Chest:
132.1 cm
142.2 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (Metric) Chest
19-295-123 2X-Large 142.2 cm
19-295-122 X-Large 132.1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-295-123 Supplier Bulwark™ FR Supplier No. SLU2LB LN XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Midweight FR Dress Uniform Shirt gives a tailored look and offers features for improved comfort and performance with CAT 2 AR/FR protection.

  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection
  • Hazard:
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 88% Cotton/12% Nylon
  • Closure: Button-down front and topstitched cuff with button closure
  • Collar: Banded, button-down collar
  • Pocket: Two chest pockets
  • Features:
    • Full underarm gussets
    • Tailored sleeve placket
    • Topstitched cuff with button closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Size 2X-Large
Gender Men's
Material Cotton, Nylon
Color Blue
Closure Type Button
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E, Category 2 Protection
Size (English) Chest 56 in.
Size (Metric) Chest 142.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.