Learn More
Bulwark™ FR Excel FR™ ComforTouch™ HRC2 Dress Uniform Shirt
Catalog No. 18120274 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
18-120-274 X-Large
1 options
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Catalog No. 18-120-274 Supplier Bulwark™ FR Supplier No. SLU2NV RG XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Arc Rating ATPV 8.6 calories/cm2
  • Flame-resistant, 7 oz. 88% cotton/12% nylon
  • Banded, topstitched, button-down collar
  • Topstitched cuffs with adjustable button closures
  • Placket front with button closure
  • Tailored sleeve placket
Specifications
Size X-Large
Material Cotton, Nylon
Color Navy