Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Midweight Excel FR™ Flame Resistant Navy Work Pant, Waist Sizes 28 to 40 in.

Catalog No. 17150406 Shop All Red Kap Industries Products
Change view
Click to view available options
Size (English) Inseam:
30 in.
31 in.
32 in.
33 in.
34 in.
36 in.
37 in.
Size:
28
29
30
31
32
33
34
35
36
38
40
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Inseam Size
17-150-406 34 in. 28
17-150-405 32 in. 28
17-150-404 30 in. 28
17-150-407 30 in. 29
17-150-408 32 in. 29
17-150-409 34 in. 29
17-150-412 32 in. 30
17-150-410 36 in. 29
17-150-413 34 in. 30
17-150-414 30 in. 31
17-150-411 30 in. 30
17-150-416 34 in. 31
17-150-417 37 in. 31
17-150-421 30 in. 33
17-150-420 34 in. 32
17-150-418 30 in. 32
17-150-419 32 in. 32
17-150-415 32 in. 31
17-150-422 32 in. 33
17-150-424 37 in. 33
17-150-423 34 in. 33
17-150-427 32 in. 34
17-150-425 30 in. 34
17-150-426 31 in. 34
17-150-429 34 in. 34
17-150-428 33 in. 34
17-150-430 30 in. 35
17-150-431 32 in. 35
17-150-434 30 in. 36
17-150-432 34 in. 35
17-150-436 32 in. 36
17-150-433 37 in. 35
17-150-435 31 in. 36
17-150-437 33 in. 36
17-150-438 34 in. 36
17-150-439 30 in. 38
17-150-440 31 in. 38
17-150-443 30 in. 40
17-150-441 32 in. 38
17-150-442 34 in. 38
17-150-444 32 in. 40
17-150-445 34 in. 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
This item is not returnable. View return policy
Catalog No. 17-150-406 Supplier Bulwark™ FR Supplier No. PEW2NV 28 34
Add to Cart
Add to Cart
This item is not returnable. View return policy

This midweight work pant offers a classic look with CAT 2 AR/FR protection and NFPA™ 2112 compliance.

  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Excel FR™ Flame resistant, 9 oz. (305 g/m2) Twill
  • Blend: 100% Cotton
  • Closure: Waistband with button closure
  • Pocket: Two slack-style front pockets and left hip pocket with button closure
  • Features:
    • Waistband with button closure
    • Two slack-style pockets
    • Left hip pocket with button closure
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 11
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size 28
Gender Men's
Product Type Flame-resistant Work Pants
Material Cotton
Color Navy
No. of Pockets 4
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating ATPV 10.6 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 34 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.