Learn More
Bulwark™ FR Excel FR™ Long Sleeve Henley HRC2 Shirt
Catalog No. 18120320 Shop All Red Kap Industries Products
Change view
Click to view available options
Gray; Regular; X-Large
Catalog No.
18-120-320 Gray; Regular; X-Large
1 options
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Catalog No. 18-120-320 Supplier Bulwark™ FR Supplier No. SEL2GY RG XL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Protection: Arc Rating EBT 11.0 calories/cm2
  • Flame-resistant 6.25 oz. interlock FR 100% cotton
  • Three button-front closure
  • FR cotton rib knit cuffs and collar
Specifications
Size X-Large (Regular)
Material FR Cotton
Color Gray
Hazard Risk Category 2
Arc Flash Rating EBT 11.0 Calories/cm2
Weight (English) 6.25 oz.