Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's Flame Resistant Long Sleeve Base Layer

Catalog No. 17200707 Shop All Red Kap Industries Products
Change view
Click to view available options
Size (English) Chest:
35 to 38 in.
38 to 41 in.
41 to 45 in.
45 to 49.5 in.
49.5 to 54 in.
54 to 58.5 in.
58.5 to 63 in.
63 to 67.5 in.
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Size (Metric) Chest:
104.14 to 114.3 cm
114.3 to 125.73 cm
125.73 to 137.16 cm
137.16 to 148.6 cm
148.6 to 160 cm
160 to 171.45 cm
89 to 96.5 cm
96.5 to 104.14 cm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Chest Size Size (Metric) Chest
17-200-707 49.5 to 54 in. 2X-Large 125.73 to 137.16 cm
17-200-702 63 to 67.5 in. 5X-Large 160 to 171.45 cm
17-200-700 54 to 58.5 in. 3X-Large 137.16 to 148.6 cm
17-200-703 41 to 45 in. Large 104.14 to 114.3 cm
17-200-704 38 to 41 in. Medium 96.5 to 104.14 cm
17-200-701 58.5 to 63 in. 4X-Large 148.6 to 160 cm
17-200-705 35 to 38 in. Small 89 to 96.5 cm
17-200-706 45 to 49.5 in. X-Large 114.3 to 125.73 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-200-707 Supplier Bulwark™ FR Supplier No. MPU8GY RG XXL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

This Long Sleeve CAT 1 AR/FR Base Layer provides an additional layer of protection within your FR layered system. Plus, the Bulwark FR logo at the collar silently signals compliance to safety managers.

  • Primary Closure: No Closure
  • Wash Care: Home Wash
  • Fabric: Excel FR™ Flame resistant, 5.5 oz. (185 g/m2)
  • Blend: 90% Cotton/10% Polyester
  • Finish: Moisture management
  • Collar: Self-fabric crewneck collar
  • Features:
    • Self-fabric crewneck collar
    • Hemmed, raglan sleeve
    • Contrast mesh full underarm gussets
    • Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 6.4
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Slim Fit
  • Gender: Male
  • Materials: Cotton/Polyester
  • Brand: Bulwark™

Specifications

Size 2X-Large
Gender Men's
Material Cotton, Polyester
Color Gray
Neck Style Tagless Crew
Sleeve Style Raglan Sleeve
Finish Ammonia Cure
Resistance Flame
Hazard Risk Category HRC1
Arc Flash Rating ATPV 6.4 Calories/cm2
Certifications/Compliance Meets NFPA, 70E Compliant
Size (English) Chest 49.5 to 54 in.
Size (Metric) Chest 125.73 to 137.16 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.