Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VF Workwear Bulwark Women's Flame Resistant Curvy Fit Denim Jeans

Catalog No. 17150630 Shop All Red Kap Industries Products
Change view
Click to view available options
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
Size:
6
8
10
12
14
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Inseam Size
17-150-630 28 in. 14
17-150-615 30 in. 6
17-150-616 32 in. 6
17-150-617 34 in. 6
17-150-618 28 in. 8
17-150-619 30 in. 8
17-150-620 32 in. 8
17-150-621 34 in. 8
17-150-622 28 in. 10
17-150-626 28 in. 12
17-150-623 30 in. 10
17-150-624 32 in. 10
17-150-625 34 in. 10
17-150-627 30 in. 12
17-150-629 34 in. 12
17-150-628 32 in. 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 17-150-630 Supplier VF Workwear Supplier No. PMJ5DD 14 28

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size 14
Gender Women's
Material Cotton, Modacrylic
Color Blue
No. of Pockets 5
Closure Type Button
Hazard Risk Category HRC2
Arc Flash Rating EBT 13.0 Calories/cm2
Size (English) Inseam 28 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.