Learn More
AdipoGen Bumetanide
SDP
Supplier:  AdipoGen AGCR13543M250
Encompass_Preferred
Chemical. CAS 28395-03-1. Formula C17H20N2O5S. MW 364.4. Bumetanide is a loop diuretic that is clinically used to treat congestive heart failure. Bumetanide also exhibits anxiolytic, neuroprotective and anti-epileptic/anticonvulsant activities. Bumetanide induces hyperpolarization in kidney cells, increasing whole-cell conductance and intracellular Ca2 levels. Bumetanide acts by antagonizing Na/K/2Cl- symporter 1 NKCC1, K/Ca2 co-transporter 2 KCC2 and NKCC2, decreasing fluid secretion and Na/K transport. While NKCC1 is expressed both in the CNS and in systemic organs, NKCC2 is kidney-specific. Bumetanide is a potent and specific inhibitor of Na-K-2Cl cotransporter 1 NKCC1 IC50 = 0.68µM. It is selective for NKCC1 over NKCC2 IC50 = 4µM. It is also an agonist of G protein-coupled receptor 35 GPR35 EC50 = 10µM and can inhibit carbonic anhydrases IX and XII.
Catalog No. 50-256-2312
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.