Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bumetanide, MP Biomedicals™

Catalog No. ICN15426305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15426305 5 g
02154263-2 250 mg
ICN15426301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15426305 Supplier MP Biomedicals, Inc Supplier No. 0215426305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bumetanide is used to probe the role of prostaglandin E2 in the regulation of chloride secretion across porcine endometrial epithelial cells.

  • A study of the effects of bumetanide application and subsequent sodium potassium chloride cotransporter (NKCC1) inhibition in cultured liver cells has indicated increased alpha-1-smooth muscle actin expression in 2-day-cultured hepatic stellate cells.
  • An investigation of lactacidosis treatment of suspended G6 glial cells with bumetanide included at 0.1 mM has shown diminished cell swelling due to lactacidosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Synonym bumetanidum

Specifications

Quantity 5 g
Melting Point 230 to 231°C
Physical Form Powder
Purity ≥99%
Molecular Weight (g/mol) 364.416
Chemical Name or Material Bumetanide
Recommended Storage Store at 2° to 8°C.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.