Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific BUN (Urea Nitrogen) UV Reagents

Catalog No. 23666163 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
300 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-163 300 mL
23-666-164 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-666-163 Supplier Pointe Scientific Supplier No. B7550300

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Certifications/Compliance CE Marked
Type Urea Nitrogen (BUN) UV Reagent Kit
Formulation NADH 0.3mM, Urease 1,500U/L, Glutamate Dehydrogenase >1500U/L, alpha-Ketoglutarate 4mM, pH 8.2 ± 0.1 Buffer, activators and non-reactive Stabilizers
For Use With (Application) Quantitative determination of Urea Nitrogen in Serum, in-vitro diagnostic use only
Quantity 300 mL
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.