Learn More
Cole-Parmer™ Bunsen Burners
Catalog No. 03391301 Shop All Oakton Products
Change view
Click to view available options
Includes:
Foot Switch
IR Sensor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03-391-301 Foot Switch
03-391-300 IR Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Safety features include monitored burn time, gas consumption, zero pressure shut off, over-heat alarm, automatic gas valve shut-off if flame extinguishes, overheating protection and burner head control that detect faulty, clogged or missing burner heads; Residual heat display illuminates as a warning to not touch the unit while the burner head is hot
Catalog No. 03-391-301 Supplier Cole-Parmer™ Supplier No. 0439718
Add to Cart
Add to Cart
Safety features include monitored burn time, gas consumption, zero pressure shut off, over-heat alarm, automatic gas valve shut-off if flame extinguishes, overheating protection and burner head control that detect faulty, clogged or missing burner heads; Residual heat display illuminates as a warning to not touch the unit while the burner head is hot
Designed with safety in mind
  • Automatic gas valve shut off if the flame extinguishes
  • Burner head detects faulty, clogged, or missing burner heads and protects against overheating
  • Residual heat display illuminates and warns users if the burner head is hot
  • Control both gas and air flow for a broad range of flame temperatures
  • Use either your house gas line or disposable gas cartridges (adapters sold separately)
These advanced Bunsen burners are compact, versatile, and designed for safety. Operate with either house gas or purchase adapters to use with disposable gas cartridges. Adjust gas and air flow with the separate control knobs to obtain a wide range of flame temperatures and intensity-great for any laboratory application. Both models feature automatic gas valve shut off should the flame extinguish, overheating protection, warning light if the head is hot, and burner head control that detects faulty, clogged or missing burner heads.
Specifications
Gas Type Butane, Natural Gas, Propane
Length (English) 12 in.
Length (Metric) 30 cm
Material Stainless Steel
Includes Foot Switch
Product Type Bunsen Burner