Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B392525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B392525G 25 g
B39255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B392525G Supplier TCI America Supplier No. B392525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18010-40-7
Molecular Formula C18H29ClN2O
Molecular Weight (g/mol) 324.893
MDL Number MFCD00078956
InChI Key SIEYLFHKZGLBNX-UHFFFAOYSA-N
Synonym 1-Butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide Hydrochloride
PubChem CID 64737
ChEBI CHEBI:77441
IUPAC Name 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
SMILES CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.Cl

Specifications

Melting Point 257°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 324.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bupivacaine Hydrochloride
RTECSNumber TK6125000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.