Learn More
Description
Bupivacaine Hydrochloride USP is a white, odorless, crystalline powder.
Alternative name(s): bupivacaine hydrochloride monohydrate, Bupivacaine HCL
Specifications
Specifications
| Chemical Name or Material | Bupivacaine Hydrochloride USP (Monohydrate) |
| Packaging | Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 73360-54-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 342.9 |
| Quantity | 25 g |
| Percent Purity | 98.5 to 101.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
