Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bupivacaine, PROMO, Thermo Scientific Chemicals
Bupivacaine is a potent and long-acting in vivo local anesthetic agent.
Supplier: Thermo Scientific Chemicals J6274214PR
Quantity | 25 g |
---|
Description
Application
- Bupivacaine has been shown to activate the AMPKα/BDNF signaling pathway in spinal astrocytes

Specifications
38396-39-3 | |
White | |
25 g | |
UN2811 | |
Soluble in water, ethanol, chloroform, and acetone. | |
CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C | |
288.44 g/mol | |
CHEBI:77431 | |
Bupivacaine |
106°C to 110°C | |
C18H28N2O | |
MFCD00243007,MFCD00941462 | |
bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine | |
LEBVLXFERQHONN-UHFFFAOYNA-N | |
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide | |
2474 | |
Crystalline Powder |
Safety and Handling
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 253-911-2
TSCA : Yes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction