Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine

Catalog No. AAJ6274206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6274206 5 g
AAJ6274203 1 g
AAJ6274214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6274206 Supplier Thermo Scientific Chemicals Supplier No. J6274206
Add to Cart

Bupivacaine, CAS # 38396-39-3, is a potent and long-acting in vivo local anesthetic agent. | CAS: 38396-39-3 | C18H28N2O | 288.44 g/mol

Bupivacaine acts as a potent and long-acting local anesthetic agent in vivo. At 100 μM, it has been reported to inhibit the uptake of dopamine in striatal synaptosomes by 47%.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Application

  • Bupivacaine has been shown to activate the AMPKα/BDNF signaling pathway in spinal astrocytes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38396-39-3
Molecular Formula C18H28N2O
Molecular Weight (g/mol) 288.44
MDL Number MFCD00243007,MFCD00941462
InChI Key LEBVLXFERQHONN-UHFFFAOYNA-N
Synonym bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine
PubChem CID 2474
ChEBI CHEBI:77431
IUPAC Name 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
SMILES CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C

Specifications

Melting Point 106°C to 110°C
Color White
Quantity 5 g
UN Number UN2811
Solubility Information Soluble in water,ethanol,chloroform (slightly ),and acetone (slightly ).
Formula Weight 288.43
Physical Form Crystalline Powder
Chemical Name or Material Bupivacaine
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 253-911-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.