Learn More
Hach Company Buret, Class A, Glass, 10 mL
Supplier:  Hach Company 2636538
Encompass_Preferred
Glass, funnel-fill buret. Certified Class A (ASTM Specification E287) with permanent serial number and certificate for NIST traceability. PTFE stopcock does not require grease. Dust cap included. 10 mL buret meets requirements for potentiometric titrations (ASTM D664).
Catalog No. 50-199-6424
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.