Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Burette Clamp, Single
SDPSCI_ED

Catalog No. S15890 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15890 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15890 Supplier United Scientific™ Supplier No. CLBUR1N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sturdy Clamp that can be used with virtually all support stands and lattice setups.

  • These clamps are constructed from aluminum and feature PVC covered spring jaws.
  • Available in single and double versions
  • These clamps accept burettes up to 35 mm O.D.
  • Clamps can be used with support rods up to 0.5 in. O.D.

Specifications

Quantity 1 Ea.
Type Burette Clamp
Material Aluminum
Color Silver
Jaw Type Spring Loaded
Diameter (Metric) Inside 35 mm max.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.