Learn More
BURTON MEDICAL LLC BURTON WOODS LAMP W/MAGNIFIER
Supplier: BURTON MEDICAL LLC UV502
5 Diopet magnifier lens with 2.25x magnification. Focal Length of 8" (20 cm). Flourescent bulbs produce UVA at approximately 365 nm wavelength. 6000 hour life . 2 UV Bulbs w/ Magnifier
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.