Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buspirone Hydrochloride, USP, 97.5-102.5%, Spectrum™ Chemical
SDP

Catalog No. 18601141 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-141 100 g Amber Glass Bottle
18-601-143 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-141 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B2850100GM

B2850, 33386-08-2, C21H31N5O2·HCl

Spectrum™ Chemical Buspirone Hydrochloride, USP is an anti-anxiety agent and a serotonin receptor agonist belonging to the azaspirodecanedione class of compounds. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 33386-08-2
Molecular Formula C21H32ClN5O2
Molecular Weight (g/mol) 421.97
InChI Key RICLFGYGYQXUFH-UHFFFAOYSA-N
IUPAC Name hydrogen 8-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-8-azaspiro[4.5]decane-7,9-dione chloride
SMILES [H+].[Cl-].O=C1CC2(CCCC2)CC(=O)N1CCCCN1CCN(CC1)C1=NC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Percent Purity 97.5 to 102.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Buspirone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.