Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butein 98.0+%, TCI America™
SDP

Catalog No. B38031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38031G 1 g
B3803100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38031G Supplier TCI America Supplier No. B38031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 487-52-5
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.256
MDL Number MFCD00017300
InChI Key AYMYWHCQALZEGT-ORCRQEGFSA-N
Synonym (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′C,3,4,4′C-Tetrahydroxychalcone
PubChem CID 5281222
ChEBI CHEBI:3237
IUPAC Name (E)-1-(2,4-dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one
SMILES C1=CC(=C(C=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O)O

Specifications

Melting Point 215°C
Quantity 1 g
Formula Weight 272.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Butein
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.