Learn More
TARGETMOL CHEMICALS INC RH1 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167401MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. RH1 (NSC 697726) is a bioreductive anticancer compound that exhibits dose-dependent biphasic effects in vitro, inducing apoptosis at higher doses and senescence at lower doses. Purity 99.15%
Catalog No. 50-287-8746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.